missing translation for 'onlineSavingsMsg'
Learn More

Creatine, TRC

Product Code. 17967454
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
1g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17967454 2.5 g 2.5g
17948314 1 g 1g
17917354 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17967454 Supplier Toronto Research Chemicals Supplier No. TRCC7814832.5G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-00-1
Molecular Formula C4 H9 N3 O2
Molecular Weight (g/mol) 131.13
Synonym Creatine (8CI), N-(Aminoiminomethyl)-N-methylglycine, 2-(1-Methylguanidino)acetic acid, Cosmocair C 100, Kre-Alkalyn, Methylguanidoacetic acid, N-Methyl-N-guanylglycine, NSC 8752, Neotine, Phosphagen, Tego Cosmo C 100, [N-(Amidino)-N-methylamino]acetic acid
IUPAC Name 2-[carbamimidoyl(methyl)amino]acetic acid
SMILES CN(CC(=O)O)C(=N)N

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C4H9N3O2/c1-7(4(5)6)2-3(8)9/h2H2,1H3,(H3,5,6)(H,8,9)
Formula Weight 131.0695
Percent Purity >95%
Chemical Name or Material Creatine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.