missing translation for 'onlineSavingsMsg'
Learn More

Creatinine, 98%

Product Code. p-7026838
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436296 10 g 10g
11446296 50 g 50g
11456296 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11436296 Supplier Thermo Scientific Alfa Aesar Supplier No. B23097.09
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 60-27-5 | C4H7N3O | 113.12 g/mol

Creatinine levels in blood and urine may be used to calculate the creatinine clearance (CrCl), which reflects the glomerular filtration rate (GFR), an important clinical index of renal function. It is a substrate for creatininase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Creatinine levels in blood and urine may be used to calculate the creatinine clearance (CrCl), which reflects the glomerular filtration rate (GFR), an important clinical index of renal function. It is a substrate for creatininase.

Solubility
Soluble in water (90 mg/ml at 20°C), alcohol (slightly), and ethanol (slightly). Insoluble in acetone, and chloroform.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-27-5
Molecular Formula C4H7N3O
Molecular Weight (g/mol) 113.12
MDL Number MFCD00059730
InChI Key DDRJAANPRJIHGJ-UHFFFAOYSA-N
Synonym creatinine, 2-imino-1-methylimidazolidin-4-one, 1-methylglycocyamidine, 1-methylhydantoin-2-imide, creatinine,heated, 4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl, 2-amino-1-methylimidazolin-4-one, creatinina, kreatinin, unii-ayi8ex34eu
PubChem CID 588
ChEBI CHEBI:16737
IUPAC Name 2-amino-3-methyl-4H-imidazol-5-one
SMILES CN1CC(=O)N=C1N

Specifications

Melting Point ∼255°C (decomposition)
pH 7.5 to 8.5
Odor Odorless
Quantity 10 g
Beilstein 112061
Merck Index 14,2569
Solubility Information Soluble in water (90mg/ml at 20°C),alcohol (slightly),and ethanol (slightly). Insoluble in acetone,and chloroform.
Formula Weight 113.12
Percent Purity 98%
Chemical Name or Material Creatinine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.