missing translation for 'onlineSavingsMsg'
Learn More

Creatinine, 98%

Product Code. 10033160 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10033160 50 g Glass bottle 50g
10553401 25 g Glass bottle 25g
11916261 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10033160 Supplier Thermo Scientific Acros Supplier No. 228940500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60-27-5 | C4H7N3O | 113.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-27-5
Molecular Formula C4H7N3O
Molecular Weight (g/mol) 113.12
InChI Key DDRJAANPRJIHGJ-UHFFFAOYSA-N
Synonym creatinine, 2-imino-1-methylimidazolidin-4-one, 1-methylglycocyamidine, 1-methylhydantoin-2-imide, creatinine,heated, 4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl, 2-amino-1-methylimidazolin-4-one, creatinina, kreatinin, unii-ayi8ex34eu
PubChem CID 588
ChEBI CHEBI:16737
IUPAC Name 2-amino-3-methyl-4H-imidazol-5-one
SMILES CN1CC(=O)N=C1N

Specifications

Melting Point 255.0°C
Color White
Flash Point 290°C
Loss on Drying 0.5% (105°C) max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 50 g
Beilstein 24,245
Merck Index 15,2557
Solubility Information (5% in water) Clear colorless to slightly yellow
Formula Weight 113.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Creatinine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.