missing translation for 'onlineSavingsMsg'
Learn More

Cresol Red, Pure, Indicator Grade, solid, Fisher Chemical

Product Code. 10755901 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10755901 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10755901 Supplier Fisher Chemical Supplier No. C/7460/44

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 290°C
CAS 1733-12-6
Molecular Formula C21H18O5S
MDL Number 5878
Formula Weight 382.42g/mol
Packaging Glass Jar
Color Green
Quantity 5 g
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.