missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cresyl Violet acetate

Product Code. 15436319
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436319 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15436319 Supplier Thermo Scientific Chemicals Supplier No. J64318.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10510-54-0 | C18H15N3O3 | 321.34 g/mol

Cresyl Violet acetate is used as stain for nerve cells and tissue spinal cord section, brain paraffin, cells and granules. It is also useful to measure pyruvate dehydrogenase complex activity when its acetylation is coupled with acetyl-CoA production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cresyl Violet acetate is used as stain for nerve cells and tissue spinal cord section, brain paraffin, cells and granules. It is also useful to measure pyruvate dehydrogenase complex activity when its acetylation is coupled with ace
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 140°C to 143°C
CAS 10510-54-0
Molecular Formula C18H15N3O3
MDL Number MFCD00013151,MFCD00013151
Synonym 9-Amino-5-imino-5H-benzo[a]phenoxazine acetate salt; Cresyl echt violet
Color Black to Green
Solubility Information Soluble in water and ethanol. Insoluble in acetone.
Quantity 10 g
InChI Key XKOCOMKJPWEOHX-UHFFFAOYSA-M
Physical Form Powder
SMILES CC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3
IUPAC Name acetic acid;5-iminobenzo[a]phenoxazin-9-amine
Molecular Weight (g/mol) 321.34
PubChem CID 44134641
Formula Weight 321.33
Chemical Name or Material Cresyl Violet acetate
Show More Show Less
compliance-icons
Product Identifier
  • Cresyl Violet acetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Cresyl Violet acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.