missing translation for 'onlineSavingsMsg'
Learn More

Cresyl violet acetate, 69%, MP Biomedicals™

Product Code. 11422112 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422112 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11422112 Supplier MP Biomedicals Supplier No. 0215072701
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Supplied as a very dark, near-black powder with a dye content of 98% for laboratory applications

Specifications

CAS 10510-54-0
Assay Percent Range 69%
Molecular Formula C18H15N3O3
MDL Number MFCD00013151,MFCD00013151
Synonym Cresyl Fast Violet, 9-amino-5-imino-5H-benzo[a]phenoxazine acetate salt
Color Green
Quantity 1 g
InChI Key XKOCOMKJPWEOHX-UHFFFAOYSA-M
Physical Form Powder
SMILES CC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3
IUPAC Name 8,12-diamino-10λ⁴-oxa-5-azatetraphen-10-ylium acetate
Molecular Weight (g/mol) 321.34
PubChem CID 44134641
Formula Weight 321.3
Grade Certified Biological Stain
Chemical Name or Material Cresyl violet acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.