missing translation for 'onlineSavingsMsg'
Learn More

Cresyl Violet acetate, pure, certified

Product Code. 10137180 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
50g
Product Code. Quantity Packaging unitSize
10137180 2.5 g Glass bottle 2.5g
10045203 10 g Glass bottle 10g
11916941 50 g Glass bottle 50g
3 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 10137180

Brand: Thermo Scientific Acros 405760025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10510-54-0 | C18H15N3O3 | 321.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10510-54-0
Molecular Formula C18H15N3O3
Molecular Weight (g/mol) 321.34
MDL Number MFCD00013151,MFCD00013151
InChI Key XKOCOMKJPWEOHX-UHFFFAOYSA-M
PubChem CID 44134641
SMILES CC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3

Specifications

Melting Point 140.0°C to 143.0°C
Absorption Ratio 0.94 to 1.2
Dye Content 65% min.
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethanol, insoluble in aceton
Infrared Spectrum Authentic
IUPAC Name acetic acid;5-iminobenzo[a]phenoxazin-9-amine
Lambda Max 596 to 599nm
Formula Weight 321.34
Grade Pure
Packaging Glass bottle
Color Black to Green
Quantity 2.5 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Cresyl Violet acetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.