missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cresyl Violet acetate, pure, high purity biological stain

Product Code. 10501481 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10501481 5 g 5g
10389033 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10501481 Supplier Thermo Scientific Chemicals Supplier No. 229630050
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10510-54-0 | C18H15N3O3 | 321.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10510-54-0
Molecular Formula C18H15N3O3
Molecular Weight (g/mol) 321.34
MDL Number MFCD00013151,MFCD00013151
InChI Key XKOCOMKJPWEOHX-UHFFFAOYSA-M
PubChem CID 44134641
IUPAC Name acetic acid;5-iminobenzo[a]phenoxazin-9-amine
SMILES CC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3

Specifications

Melting Point 140.0°C to 143.0°C
Dye Content 65% min.
Infrared Spectrum Authentic
Loss on Drying 20% max. (50°C) (0.05mm Hg)
Packaging Glass bottle
Color Black to Green
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethanol, insoluble in aceton
Quantity 5 g
Physical Form Fine Crystalline Powder
Formula Weight 321.34
Grade Pure
Chemical Name or Material Cresyl violet acetate
Show More Show Less
compliance-icons
Product Identifier
  • Cresyl Violet acetate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.