missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ CRF-1 - Controlled Rate Freezer

Product Code. 16389346
Change view
Click to view available options
Model:
CRF-1 H01
CRF-1 H05
CRF-1 H06
Holds:
Deep Cryovials (55 x 1.8 mL)
IMV Straws (18 x 0.3 mL)
SBS Microplate (1)
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Holds Unit Size
16389346 CRF-1 H01 IMV Straws (18 x 0.3 mL) Each
15982741 CRF-1 H05 SBS Microplate (1) Each
16369646 CRF-1 H06 Deep Cryovials (55 x 1.8 mL) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16389346 Supplier Grant Instruments™ Supplier No. CRF1 H01

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CRF-1 Liquid nitrogen free and cryogen free controlled rate freezer for research into the cryopreservation of a wide range of material including: embryos, stem cells, mammalian cells, spermatozoa, antibodies, tissue sections and rodent organs.

CRF-1 Liquid nitrogen free and cryogen free controlled rate freezer for research into the cryopreservation of a wide range of material including: embryos, stem cells, mammalian cells, spermatozoa, antibodies, tissue sections and rodent organs. The CRF-1 brings accuracy, precision and reproducibility to biological cryopreservation.

Unlike conventional liquid nitrogen based controlled rate cooling equipment, the CRF-1 poses no contamination risk and can be used in cleanrooms and barrier facilities. The CRF1 fits neatly and quietly on a bench-top and its performance in terms of cell viability after freezing is comparable or better than standard liquid nitrogen freezers. As alcohol is not used, there is also no potential fire risk. The CRF-1 will cool down to -100°C with straws.

The cooling rate of the CRF-1 is precisely controlled, ensuring accuracy and reproducibility throughout the freezing profile, especially for the important nucleation/seeding phase. This ensures optimal recovery of cells upon thawing. Operation is simple and can be carried out with or without a PC; data can be logged via PC software and cooling profiles are directly displayed. Different cooling profiles are available from a drop down menu and customised profiles can be written.

Main applications:
The CRF-1 is highly versatile and can be used for the cryopreservation research of a wide range
of samples in cryovials, straws, bags, microplates and Matrix-96-well block plates in the following
areas:
• Transgenic embryos research
• Stem cell research
• Clinical and research samples, e.g. lymphocytes and tissue cell lines in conventional cryovials
• Various mammalian cells including cardiomyocytes, adipose, liver and muscle
• Cord blood derived stem cells
• Adherent cells and stem cells in microplates
• Cell suspensions in numbered/barcoded arrays
• Robotic integration – the CRF-1 has also successfully been integrated into robotic systems
• Suitable for applications in veterinarian IVF

Key benefits and features:
• Accurate and reproducible control of cooling rates and sample temperatures
• Easy to use and samples can be nucleated/seeded in-situ
• Linear and non-linear cooling profiles
• Low running costs: estimated at 1% of liquid nitrogen-controlled rate freezing
• Temperature remains at -100°C at the end of cycle for straw applications until freezer
is switched off
• Un-interruptible Power Supply (UPS): complete cycle run if power fails (supplied as an
optional accessory)
• CE marked (laboratory use)
• Servicing and calibration available
• 3 year warranty

Dimensions (w x d x h): 264 x 367 x 445 mm
Weight (Kg): 9.8
Max plate temperature: 30°C
Min plate temperature: -80°C or -100°C (dependent on model)
Temperature accuracy: ± 0.5°C
Temperature stability: ± 0.1ºC
Temperature uniformity: ± 1.0°C
Max cooling rate: 10C°C /min
Max temperature stabilization time: 5 minutes

TRUSTED_SUSTAINABILITY

Specifications

Type CRF-1 H01 Controlled rate freezer for 18 x 0.3 mL IMV straws.
Model CRF-1 H01
Weight (Metric) 34 kg
Holds IMV Straws (18 x 0.3 mL)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.