missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ CRF-1 Controlled Rate Freezer

Product Code. 17285146 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Holds:
16 x 0.5 mL CBS High Security Straws
55 x 1.8 mL Cryovials (0.5 mL Max. Fill)
Flat Plate for Various Items or Vessels
Plate for 10 x 0.5 mL CBS High Security Straws and 12 x 1.8 mL Cryovials (0.5 mL Max. Fill)
Plate for 12 x 0.3 mL IMV Straws and 12 x 1.8 mL Cryovials (0.5 mL Max. Fill)
Temperature Range:
-100°C min.
-80°C min.
Packungsgröße:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Holds Temperature Range Packungsgröße
17285146 Plate for 12 x 0.3 mL IMV Straws and 12 x 1.8 mL Cryovials (0.5 mL Max. Fill) -100°C min. Each
17245146 16 x 0.5 mL CBS High Security Straws -100°C min. Each
17255146 55 x 1.8 mL Cryovials (0.5 mL Max. Fill) -80°C min. Each
17275146 Plate for 10 x 0.5 mL CBS High Security Straws and 12 x 1.8 mL Cryovials (0.5 mL Max. Fill) -100°C min. Each
17265146 Flat Plate for Various Items or Vessels -100°C min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17285146

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This liquid nitrogen-free and cryogen-free controlled rate freezer is ideal for research into the cryopreservation of a wide range of materials, including embryos, stem cells, mammalian cells, spermatozoa, antibodies, tissue sections and rodent organs.

Unlike conventional liquid nitrogen-based controlled rate cooling equipment, the CRF-1 poses no contamination risk and can be used in cleanrooms and barrier facilities. The CRF1 fits neatly and quietly on a bench-top and its performance in terms of cell viability after freezing is comparable or better than standard liquid nitrogen freezers. As alcohol is not used, there is also no potential fire risk. The CRF-1 will cool down to -100 ̊C with straws.

The cooling rate of the CRF-1 is precisely controlled, ensuring accuracy and reproducibility throughout the freezing profile, especially for the important nucleation or seeding phase. This ensures optimal recovery of cells upon thawing. Operation is simple and can be carried out with or without a PC; data can be logged via PC software and cooling profiles are directly displayed. Different cooling profiles are available from a drop down menu and customised profiles can be written.

The CRF-1 is highly versatile and can be used for the cryopreservation research of a wide range of samples in cryovials, straws, bags, microplates and Matrix-96-well block plates in the following areas:

  • Transgenic embryos research
  • Stem cell research
  • Clinical and research samples, e.g. lymphocytes and tissue cell lines in conventional cryovials
  • Various mammalian cells including cardiomyocytes, adipose, liver and muscle
  • Cord blood derived stem cells
  • Adherent cells and stem cells in microplates
  • Cell suspensions in numbered/barcoded arrays
  • Robotic integration – the CRF-1 has also successfully been integrated into robotic systems
  • Suitable for applications in veterinarian IVF

Key benefits and features:

  • Accurate and reproducible control of cooling rates and sample temperatures
  • Easy to use and samples can be nucleated/seeded in-situ
  • Linear and non-linear cooling profiles
  • Low running costs: estimated at 1% of liquid nitrogen controlled rate freezing
  • Temperature remains at -100 ̊C at the end of cycle for straw applications until freezer is switched off
  • Uninterruptible Power Supply (UPS): complete cycle run if power fails (supplied as an optional accessory)
  • CE marked (laboratory use)
  • Servicing and calibration available

Specifications:

  • A number of fixed plates available (standard straws, high security straws, cryovials, cryocyte bags and microplates)
  • Freezer can be run as a stand alone pre-programmed unit or connected to a control PC (computer with serial port and control software and Windows)
  • Software allowing configuration of existing profiles, creation of new customer profiles, data logging and reporting (all data is logged in an encrypted and Excel-readable form)
  • Standard, pre-set cooling profiles available
  • Backup electrical supply: 750 VA UPS capable of running freezer for 3 hour cycle in the case of an electrical power failure
  • Seamless with no dirt traps; may be cleaned using standard wipes
TRUSTED_SUSTAINABILITY

Spezifikation

Type Cryogenic Freezer
Temperature Range -100°C min.
Chamber Material Aluminum
Certifications/Compliance RoHS, CE, UKCA, RoHS, CE, UKCA
Style Benchtop
Color Gray
Warranty 3 Years
Defrost Automatic
Dimensions (DxWxH) Exterior 264 x 367 x 445 mm
PC Programmability CRF-1 User Software
Amperage 2.5 A
Plug Type EU, UK, US
Electrical Requirements 100/240 V (Mains Supply), 24 V DC (Unit Supply)
Frequency 50/60 Hz
Wattage 150 W
Compressor CFC-free
Weight (English) 21.6 lb.
Weight (Metric) 9.8 kg
Holds Plate for 12 x 0.3 mL IMV Straws and 12 x 1.8 mL Cryovials (0.5 mL Max. Fill)
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.