Description
The quality of a crimp seal is strongly dependent on the right calibration of the crimping tool used to apply the closure to the vial. A too tight or too loose crimp will impair the quality of the seal. To maintain good results, make sure to re-calibrate your crimping tool regularly and check the quality of the seal every time you crimp.
- Packed in zip-lock PE-bags with tear-off foil strip to ensure tamper-evidence and re-closeability.
Specifications
Specifications
| Color | Silver |
| Material | Red Rubber, White PTFE |
| Includes | With liner |
| Closure Type | Open Top Cap |
| Closure Size | ND20 |