missing translation for 'onlineSavingsMsg'
Learn More

Crocetin, TRC

Product Code. 30003693
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003693 10 mg 10mg
30005488 100 mg 100mg
30012163 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003693 Supplier Toronto Research Chemicals Supplier No. TRCC79495010MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27876-94-4
Molecular Formula C20 H24 O4
Molecular Weight (g/mol) 328.4
Synonym 2,4,6,8,10,12,14-Hexadecaheptaenedioic acid, 2,6,11,15-tetramethyl-, (all-E)- (8CI), 8,8'-Diapo-ψ,ψ-carotenedioic acid, Crocetin (6CI), (2E,4E,6E,8E,10E,12E,14E)-2,6,11,15-Tetramethyl-2,4,6,8,10,12,14-hexadecaheptaenedioic acid, Apocarotenic acid, Crocetin (stable), NSC 407300, Stable crocetin, Transcrocetin, trans-Crocetin, α-Crocetin
IUPAC Name (2E,4E,6E,8E,10E,12E,14E)-2,6,11,15-tetramethylhexadeca-2,4,6,8,10,12,14-heptaenedioic acid
SMILES C\C(=C/C=C/C=C(\C)/C=C/C=C(\C)/C(=O)O)\C=C\C=C(/C)\C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H24O4/c1-15(11-7-13-17(3)19(21)22)9-5-6-10-16(2)12-8-14-18(4)20(23)24/h5-14H,1-4H3,(H,21,22)(H,23,24)/b6-5+,11-7+,12-8+,15-9+,16-10+,17-13+,18-14+
Formula Weight 328.1675
Chemical Name or Material trans-Crocetin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.