missing translation for 'onlineSavingsMsg'
Learn More

Croconic acid, 98%

Product Code. 11434837
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434837 5 g 5g
11424837 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434837 Supplier Thermo Scientific Alfa Aesar Supplier No. B21809.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 488-86-8 | C5H2O5 | 142.066 g/mol

Croconic acid is used to prepare 1,3-bis-(2-dimethylamino-5-thienyl)croconine by reacting with dimethyl-thiophen-2-yl-amine. It is also used in organic synthesis and employed as a dye for biological research purposes. Further, it is involved in the preparation of ethers such as dimethyl croconate. In addition to this, it acts as a ligand and form coordination complexes with metals such as barium, copper, silver and lead.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Croconic acid is used to prepare 1,3-bis-(2-dimethylamino-5-thienyl)croconine by reacting with dimethyl-thiophen-2-yl-amine. It is also used in organic synthesis and employed as a dye for biological research purposes. Further, it is involved in the preparation of ethers such as dimethyl croconate. In addition to this, it acts as a ligand and form coordination complexes with metals such as barium, copper, silver and lead.

Solubility
Soluble in water and ethanol.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 488-86-8
Molecular Formula C5H2O5
Molecular Weight (g/mol) 142.066
MDL Number MFCD00181389
InChI Key RBSLJAJQOVYTRQ-UHFFFAOYSA-N
Synonym croconic acid, crocic acid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione, unii-6hgy35uxaa, 6hgy35uxaa, 4-cyclopentene-1,2,3-trione, 4,5-dihydroxy, croconicacid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione #
PubChem CID 546874
IUPAC Name 4,5-dihydroxycyclopent-4-ene-1,2,3-trione
SMILES C1(=C(C(=O)C(=O)C1=O)O)O

Specifications

Melting Point ∼280°C (decomposition)
Odor Odorless
Quantity 5 g
Solubility Information Soluble in water and ethanol.
Formula Weight 142.07
Percent Purity 98%
Chemical Name or Material Croconic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.