missing translation for 'onlineSavingsMsg'
Learn More

Cromolyn sodium salt, 98%

Product Code. 15436739
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436739 1 g 1g
15446739 5 g 5g
15456739 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15436739 Supplier Thermo Scientific Alfa Aesar Supplier No. J65122.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mast cell stabilizer that prevents release of histamine from mast cells | CAS: 15826-37-6 | C23H14Na2O11 | 512.33 g/mol

Cromolyn sodium salt is used as a mast cell stabilizer that prevents release of histamine and other pro-inflammatory mediators from mast cells. Cromolyn sodium salt is used as a chemopreventive and an anti-inflammatory agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cromolyn sodium salt is used as a mast cell stabilizer that prevents release of histamine and other pro-inflammatory mediators from mast cells. Cromolyn sodium salt is used as a chemopreventive and an anti-inflammatory agent.

Solubility
Soluble in water to 100mg/ml

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15826-37-6
Molecular Formula C23H14Na2O11
Molecular Weight (g/mol) 512.33
MDL Number MFCD00057744
InChI Key VLARUOGDXDTHEH-UHFFFAOYSA-L
Synonym cromolyn sodium, sodium cromoglycate, disodium cromoglycate, sodium cromoglicate, sodium cromolyn, gastrocrom, nasalcrom, cromoglycate disodium, natrium cromoglicat, lomudal
PubChem CID 27503
ChEBI CHEBI:128458
SMILES [Na+].[Na+].OC(COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O)COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O

Specifications

Color White to Yellow
Quantity 1 g
Merck Index 14,2590
Solubility Information Soluble in water to 100mg/ml
IUPAC Name disodium;5-[3-(2-carboxylato-4-oxochromen-5-yl)oxy-2-hydroxypropoxy]-4-oxochromene-2-carboxylate
Formula Weight 512.33
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Cromolyn sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.