Learn More
Thermo Scientific™ Crossflow Filtration Accessory Pack
Product Code. 18031663 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Each
Unit Size:
Each
Product Code. Quantity unitSize
18031663 Each Each
1 options
Product Code. 18031663 Supplier Thermo Scientific™ Supplier No. A57937
  • ONLINE EXCLUSIVE PRICE
    49.27 €
    42.86 € / Pack of 1
    Save 6.41 €
    13% Off
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
The Crossflow Filtration Accessory Pack features all necessary components including the feed line, return line, filtrate line, and pressure gauge.
Accessory pack includes:
• Feed line
• Return line
• Filtrate line
• Pressure gauge
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Disposable Reusable
Material PVC, Polypropylene, Stainless Steel
Sterility Non-sterile
Includes Feed line, return line, filtrate line, and pressure gauge
Quantity Each
Fit for Purpose Certified Quality
For Use With (Equipment) Filtration Systems