missing translation for 'onlineSavingsMsg'
Learn More

Notrax Crossrib Sof-Tred™ Dyna-Shield™ Layer with Side-to-Side Wide Ribs, Gray

Product Code. 17722058
Change view
Click to view available options
Dimensions (L x W):
91 x 60 cm
150 x 91 cm
1830 x 60 cm
1830 x 91 cm
1830 x 122 cm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
17722058 150 x 91 cm Each
17702058 91 x 60 cm Each
17742048 1830 x 60 cm Each
17762048 1830 x 91 cm Each
17782048 1830 x 122 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17722058 Supplier Notrax Supplier No. 406S0035GY
  • ONLINE EXCLUSIVE PRICE
    157.67 €
    128.06 € / Pack of 1
    Save 29.61 €
    18% Off
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Notrax Crossrib Sof-Tred™ - Exceptional worker comfort due to 12.7 mm thick specially formulated compound. Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl.

  • Exceptional worker comfort due to 127 mm thick specially formulated compound
  • Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl supported by independent laboratory testing
  • Side-to-side wide ribs provide traction and easy to sweep clean
  • Available with yellow borders along sides to comply with OSHA code 1910-144
  • All 4 sides are bevelled and sealed to minimize tripping hazards

Specifications

Dimensions (L x W) 150 x 91 cm
Color Gray
Length (English) 59 in.
Length (Metric) 150 cm
Material Microcellular Vinyl
Type Industrial and Commercial Matting
Width (Metric) 91 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.