missing translation for 'onlineSavingsMsg'
Learn More

Tempshield™ Cryo-Gloves™ Blue Elbow Length Cryogenic Gloves

Product Code. 15878542
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
1 pair
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15878542 Small 1 pair
11390530 Medium 1 pair
11883811 Large 1 pair
16082222 X-Large 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.
Product Code. 15878542 Supplier Tempshield™ Supplier No. EBS
  • ONLINE EXCLUSIVE PRICE
    244.00 €
    187.34 € / 1 pairs
    Save 56.66 €
    23% Off
View all promotions
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.

State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).

State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).

  • Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
  • State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
  • High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
  • Ideal for applications where function is important and safety is critical
  • Not intended for immersion in liquid nitrogen or other cryogenic liquids

Specifications

Certifications/Compliance (Europe) EN 420, EN 511
Description Elbow Length
Color Blue
Hand Specific
Product Type Cryogenic Gloves
Size Small
EN388 Standard Yes
For Use With (Application) Dry Ice Handling, Low/Ultra-Low Temperature Freezers, and Closed Cryogenic Systems
Series Cryo-Gloves™
Temperature (English) Min -321°F
Temperature (Metric) Min. -196°C
Length (English) 17.3 to 19.8 in.
Length (Metric) 44 to 50 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.