Learn More
Scilabub™ Frosters™ Cryogenic Mid-Arm Glove
Product Code. 11893831
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
1 pair
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11893831 Large 1 pair
11883831 Medium 1 pair
11883801 Small 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11893831 Supplier Scilabub™ Supplier No. GLO/CML
  • ONLINE EXCLUSIVE PRICE
    367.42 €
    180.41 € / 1 pairs
    Save 187.01 €
    50% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Frosters™ Polyimide Cryogenic Gloves have been specially developed for the handling of very cold objects, typically for objects stored in the vapour phase of liquid nitrogen (ranging from -120°C to -190°C).
The middle layer is of special polyolefin/polyester micro-fibre, which is almost twice as effective in thermal insulation as a similar thickness of down. This minimises heat transfer without being so thick as to be cumbersome. The inner layer is of cotton for comfortable wear. They are made to EN 420 sizes and have been tested to EN388 (2, X, 2, X, ) and EN 511-contact cold 2, convective cold 2, Flexibility PASS. Avoid contact with alcohol-based products. To reduce bulk around the fingers, the stitch holes have not been sealed. In normal use this has not proved a problem, however, the gloves should not be immersed in liquid nitrogen and it is not advisable to use them above -80°C. Very durable, they retain their flexibility (albeit with some stiffening) for long periods in cold atmospheres. Polyimide outer material, polyolefin with some polyester as filling and cotton liner. Manufactured to EN511.Developed for handling very cold objects, typically those stored in the vapour phase of liquid nitrogen (in the range -120°C to -190°C).
Specifications
Color Blue
Material Fabric
Powdered Powder Free
Product Type Cryogenic Gloves
Size Large
Sterility Non-sterile
Quantity 1 Pack
Resistant To Water