Description
- For safe and convenient removal of frozen vials from storage boxes
- Simply insert picker into notch located in top of cryoclear caps and pull vial out
Specifications
Specifications
| Color | Blue |
| Sterility | Non-sterile |