Learn More
Thermo Scientific™ CryoMed™ Controlled-Rate Freezer IVF Accessories
Product Code. 11869480 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Cane Holder
Straw Holder
Vial Holder
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11869480 Cane Holder Each
10795505 Straw Holder Each
11859480 Vial Holder Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11869480 Supplier Thermo Scientific™ Supplier No. 4000714
  • ONLINE EXCLUSIVE PRICE
    133.00 €
    127.00 € / Pack of 1
    Save 6.00 €
    4% Off
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
Integrate Thermo Scientific™ CryoMed™ Controlled-Rate Freezer IVF Accessories with your current system for increased functionality.
For use in CryoMed 7456, 7457, 7475, 7458, 7459, and 7476 freezers.
TRUSTED_SUSTAINABILITY
Specifications
Description for 1.2 and 2.0 vials
Type Cane Holder
For Use With (Equipment) All CryoMed Controlled-Rate Freezers
Holds Cane