missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CryoMed™ with OPC UA, General Purpose
BIOPHARMA

Product Code. 17445173 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
17 L
34 L
48 L
Type:
Controlled-rate Freezer with Printer
Controlled-rate Freezer without Printer
Electrical Requirements:
220-230 V, 50-60 Hz
220-230V, 50-60Hz
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Type Electrical Requirements Unit Size
17445173 34 L Controlled-rate Freezer with Printer 220-230V, 50-60Hz Each
17535865 34 L Controlled-rate Freezer without Printer 220-230 V, 50-60 Hz Each
17555865 48 L Controlled-rate Freezer without Printer 220-230V, 50-60Hz Each
17515865 17 L Controlled-rate Freezer with Printer 220-230V, 50-60Hz Each
17525865 17 L Controlled-rate Freezer without Printer 220-230V, 50-60Hz Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17445173 Supplier Thermo Scientific™ Supplier No. TSCM34FV
  • ONLINE EXCLUSIVE PRICE
    46670.23 €
    42500.00 € / Pack of 1
    Save 4170.23 €
    8% Off
View all promotions
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific CryoMed Controlled-Rate Freezers with OPC UA are designed with reliability, protection, and data traceability in mind for your sample preparation and GMP production needs.

For customers in cell and gene therapy and vaccine production, the CryoMed with OPC UA provides precise, repeatable freezing results that protect samples from intracellular freezing. The CryoMed features OPC UA serial communication (ethernet) capabilities, comes standard factory certifications, provides enhanced data traceability via a touch screen user interface and offers customizable freezing profiles while supporting 21 CFR part 11 and GMP needs.

User-friendly operation with enhanced data traceability

  • Intuitive touchscreen display allows for easy set-up, operation, and review of a freezing run
  • OPC UA (ethernet) serial communication protocol
  • Factory-issued certificates
  • Six pre-set freezing profiles and space for up to 14 user-defined, 'custom' freeze profiles
  • Integrated touchscreen UI logs usage and events to support 21 CFR part 11 requirements
  • USB data export of .pdf file run logs support 21 CFR part 11
  • User security: 3 levels of user accessibility
  • 'Run last' feature allows the same profile to be run consecutively with the touch of a button
  • PC interface software provides remote operation capabilities, run review, and custom profile creation

Real-time run monitoring for sample protection

  • Chamber and sample temperatures are monitored by Type T thermocouples, eliminating lag time and providing 'real time' responsiveness
  • Standard alarms to alert users of thermocouple failures, heater malfunction, high/low temperature limits, temperature tracking, power failure, and completion of run
  • Remote alarm contacts
  • Optional thermal printer

Reliable temperature performance for high throughput usage

  • Dual solenoid valves are designed to balance LN2 injection volume for precise temperature control and accelerated freezing
  • Preventative maintenance indicator for LN2 solenoid replacement limits downtime
  • Temperature control and uniformity achieved via an air-handling and liquid nitrogen injection ring system
  • Compatible with VHP cleaning techniques
  • Environmentally friendly insulating foam
  • Type 304 stainless steel with exterior powder-coat finish

Warranty varies by region. Check with your local representative for more information.

TRUSTED_SUSTAINABILITY

Specifications

Type Controlled-rate Freezer with Printer
Capacity (Metric) 34 L
Temperature Range -180°C to +50°C
No. of Doors 1
Cabinet Material Stainless Steel
Exterior Finish Powder-coated steel
Certifications/Compliance CE
Style Freestanding
Display Touchscreen user interface
Warranty Warranty varies by region. Contact your local sales representative for more information.
Sample Loading Front-loading
Dimensions (DxWxH) Exterior 24.3 x 43.3 x 21.7 in. (61 x 109.9 x 55.1 cm)
Dimensions (D x W x H) Interior 13 x 13 x 12 in. (33 x 33 x 30.5 cm)
Monitoring Options Alarms, Remote Alarm Contacts
Data Outputs USB and OPC UA (ethernet)
On-Board Datalogging Yes
PC Programmability PC Interface software supported
Connectivity Thermo Fisher Connect compatible
Amperage 5.5A
Plug Type CEE 7/7
Voltage 208/230 V
Electrical Requirements 220-230V, 50-60Hz
Vial Capacity <800
Compressor Air-cooled
Weight (English) 174 lb.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.