Learn More
Thermo Scientific™ CryoMed™ Sensors
Product Code. 10590275 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
1.2/2.0 Cryo Tubes in CryoMed
4.0/5.0 Cryo Tubes in CryoMed
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10590275 4.0/5.0 Cryo Tubes in CryoMed Each
10061204 1.2/2.0 Cryo Tubes in CryoMed Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10590275 Supplier Thermo Scientific™ Supplier No. 4000386
  • ONLINE EXCLUSIVE PRICE
    585.00 €
    569.00 € / Pack of 1
    Save 16.00 €
    2% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Improve the functionality of Thermo Scientific™ Cryomed™ controlled-rate freezers with these CryoMed Sensors.
Features:
  • Thermocouple sensors for cryo tubes, sheath straw and vial samples
  • Includes male plug
Recommended for:
Controlled-rate freezing applications
TRUSTED_SUSTAINABILITY
Specifications
Description Thermocouple Sensor
Type Thermocouple Sensor
Capacity (Metric) 5 mL
For Use With (Equipment) 4.0/5.0 Cryo Tubes in CryoMed
Plug Type Male
Holds Cryo Tube, Sheath Straw, Vial