Learn More
Taylor Wharton™ Cryorack Box
Product Code. p-8010211
Click to view available options
Holds:
100 Tubes
Unit Size:
Each
Product Code. 11798409
Brand: Taylor Wharton™ TKW1
Only null left
Add to basket
Edge
Add to basket
The aluminum Cryorack Box can withstand the very coldest temperatures and are for use with the K Series Cryogenic Storage System.
  • Square rack for cryobox tubes
  • To be mixed according to diagram opposite to optimize storage
Specifications
Holds 100 Tubes
Material Aluminum
Type Cryorack Box