missing translation for 'onlineSavingsMsg'
Learn More

Cryptotanshinone, MedChemExpress

Product Code. 16438888
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16438888 10 mg 10mg
16464777 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16438888 Supplier Medchem Express Supplier No. HYN0174

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cryptotanshinone is a natural compound extracted from the root of Salvia miltiorrhiza Bunge that shows antitumor activities. Cryptotanshinone inhibits STAT3 with an IC50 of 4.6 μM.

Cryptotanshinone is a natural compound extracted from the root of Salvia miltiorrhiza Bunge that shows antitumor activities. Cryptotanshinone inhibits STAT3 with an IC50 of 4.6 μM.

Specifications

Chemical Name or Material Cryptotanshinone
CAS 35825-57-1
Color Orange
Purity Grade Notes Research
Molecular Formula C19H20O3
Quantity 10 mg
Synonym Cryptotanshinon Tanshinone c
Solubility Information DMSO : 2 mg/mL (6.75 mM; ultrasonic and warming and heat to 80°C) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=C2C=CC3=C1CCCC3(C)C)C(C4=C2OC[C@@H]4C)=O
Molecular Weight (g/mol) 296.36
Formula Weight 296.36
Percent Purity 98.02%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.