missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Crystalline Peptide
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Purification Method | HPLC |
| Content And Storage | -30°C to -10°C |
| Format | Solid |
| Molecular Weight (g/mol) | 261.28 |
| Product Type | Crystalline Peptide |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
| Sequence | Trp-Gly |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction