missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CSF1 Split FISH Probe

Product Code. 16093525
Change view
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16093525 200 μL 200µL
16083525 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16093525 Supplier Abnova™ Supplier No. FS0071.200uL
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe

Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.

Specifications

Content And Storage Store at 4°C in the dark.
Description DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Format Tube
For Use With (Application) FISH-Ce
Gene ID (Entrez) 1435
Includes DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Quantity 200 μL
Target Human
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.