missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ CSM-His-Leu-Lys-Trp-Ura

Product Code. 11347939 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347939 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11347939 Supplier MP Biomedicals™ Supplier No. 114550022
  • ONLINE EXCLUSIVE PRICE
    1005.00 €
    950.25 € / 100g
    Save 54.75 €
    5% Off
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

CSM-His-Leu-Lys-Trp-Ura, Powder

  • 5 dropout of Complete Supplement Mixture (CSM) of amino acids for S. cerevisiae growth media.
  • Time tested formulation is available in complete and dropout formulations
  • For use with DOB or YNB with the appropriate nitrogen and carbon sources to custom formulate your own selective Synthetic Defined yeast media
  • 5 Dropout Complete Supplement Mixture used to select for yeast containing plasmids with specific nutritional markers and to select for gene function in transformation experiments
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular Biology Certified
Content And Storage Store CSM-HIS-LEU-LYS-TRP-URA powder at ambient temperature (15-30°C). CSM-HIS-LEU-LYS-TRP-URA powder is hygroscopic, therefore minimize exposure to air to prolong storage of powder mixture. Store sterile agar medium at 4°C and liquid medium at 15-30°C in subdued light.
Description 5 dropout of Complete Supplement Mixture (CSM) of amino acids for S. cerevisiaegrowth media. Time tested formulation is available in complete and dropout formulations.
Form Powder
Product Type Cell Culture Media
Sterility Sterile
Formulation Drop-Out Supplementsmg/L Adenine 0 L-Arginine HCl 50 L-Aspartic Acid 80 L-Histidine HCl 0 L-Isoleucine 50 L-Leucine 0 L-Lysine HCl 50 L-Methionine 20 L-Phenylalanine 50 L-Threonine 100 L-Tryptophan 0 L-Tyrosine 50 Uracil 0 L-Valine 140
For Use With (Application) Used for S. cerevisiae growth media
pH 4.0 to 4.4
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.