missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ CSM-Met-Ura

Product Code. 11377919 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377919 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11377919 Supplier MP Biomedicals™ Supplier No. 114520822

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CSM-Met-Ura, Powder

  • 5 dropout of Complete Supplement Mixture (CSM) of amino acids for S. cerevisiae growth media.
  • Time tested formulation is available in complete and dropout formulations
  • For use with DOB or YNB with the appropriate nitrogen and carbon sources to custom formulate your own selective Synthetic Defined yeast media
  • Double Dropout Complete Supplement Mixture used to select for yeast containing plasmids with specific nutritional markers and to select for gene function in transformation experiments
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular Biology Certified
Content And Storage Store CSM-Met-Ura powder at ambient temperature (15-30°C). CSM powder is hygroscopic, therefore minimize exposure to air to prolong storage of powder mixture. Store sterile agar medium at 4°C. And liquid medium at 15-30°C in subdued light.
Form Powder
Product Type Cell Culture Media
Sterility Sterile
Sterilization Method Autoclave at 121°C for 25 min.
Formulation Adenine 10mg/L; L-Arginine HCl 50mg/L; L-Aspartic Acid 80mg/L; L-Histidine HCl 20mg/L; L-Isoleucine 50mg/L; L-Leucine 100mg/L; L-Lysine HCl 50mg/L; L-Methionine 0mg/L; L-Phenylalanine 50mg/L; L-Threonine 100mg/L; L-Tryptophan 50mg/L; L-Tyrosine 50mg/L; Uracil 0mg/L; L-Valine 140mg/L;
For Use With (Application) Used for S. cerevisiae growth media
pH Solution pH: 4.24 at 25°C
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.