missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ CSM-Trp with 20mg/L ADE (Powder)

Product Code. 11317909 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317909 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11317909 Supplier MP Biomedicals™ Supplier No. 114512522

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Time tested formulation. MP Biomedicals CSM-Trp with 20 mg/L Ade (Powder) is a single dropout of Complete Supplement Mixture of amino acids for S. cerevisiae growth media. Suitable for use with yeast cell culture, gene expression and transcription regulat

To use, add 0.76g of CSM-Trp+Ade to each liter of purified water and DOB or DOBA. Then, autoclave at 121°C for 25 minutes for complete synthetic defined single drop-out medium for S.cerevisiae.

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
For Use With (Application) Yeast cell culture, Gene Expression, Transcription Regulatiotn Research
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.