missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CSPD™ Substrate (0.4 mM Ready-To-Use) with Emerald-II™ Enhancer, Size A

Product Code. 15894973
Change view
Click to view available options
Quantity:
100 ml
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15894973 100 ml 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15894973 Supplier Invitrogen™ Supplier No. T2212

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ready-to-use chemiluminescent alkaline phosphatase substrate, for protein or nucleic acid blotting on nitrocellulose membranes or solution-based assays.

Chemiluminescent substrates provide highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP). Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.

  • Detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed and ease
  • Exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting
  • Use in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays
  • Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours
  • Enhancers increase light output and sensitivity
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Substrate
Quantity 100 ml
Detection Method Chemiluminescence
Form Liquid
Format Bottle
Product Line CSPD, NovaBright
Substrate Type Alkaline Phosphatase
Content And Storage Store the product at 4°C.
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.