missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ CSPD™ Substrate (25 mM Concentrate)

Product Code. 12664315
Change view
Click to view available options
Quantity:
0.5 mL
1 mL
2 mL
5 mL
10 mL
25 mL
100 mL
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12664315 1 mL Each
12614315 0.5 mL Each
12634315 2 mL Each
12624315 5 mL Each
12654315 25 mL Each
12624325 100 mL Each
12644315 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 12664315 Supplier Molecular Probes™ Supplier No. T2098
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

This concentrated CSPD™ substrate is a chemiluminescent alkaline phosphatase substrate for protein or nucleic acid blotting on nitrocellulose membranes. This substrate can also be used in solution-based assays.

This CSPD™ chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.

• Chemiluminescent substrates provide highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP).
• Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.

This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours. Solution assays require chemiluminescent enhancers. Membrane-based assays may also require them for increased light output and sensitivity.

For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the product at 4°C.
Form Liquid
For Use With (Application) Southern, Northern, and Western Blotting
Product Line CSPD
Quantity 1 mL
Substrate Alkaline Phosphatase
Substrate Type AP (Alkaline Phosphatase) Substrate
Format Bottle(s)
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.