missing translation for 'onlineSavingsMsg'
Learn More

CTEP, MedChemExpress

Product Code. 16489107
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489107 5 mg 5mg
16482617 50 mg 50mg
16426528 10 mg 10mg
16406776 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16489107 Supplier Medchem Express Supplier No. HY15445
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress CTEP (RO 4956371) is a novel, long-acting, orally bioavailable allosteric antagonist of mGlu5 receptor with IC50 of 2.2 nM, and shows > 1000-fold selectivity over other mGlu receptors.

CTEP (RO 4956371) is a novel, long-acting, orally bioavailable allosteric antagonist of mGlu5 receptor with IC50 of 2.2 nM, and shows > 1000-fold selectivity over other mGlu receptors.

Specifications

Chemical Name or Material CTEP
CAS 871362-31-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H13ClF3N3O
Quantity 5 mg
Synonym RO 4956371 mGluR5 inhibitor
Solubility Information DMSO : ≥ 100 mg/mL (255.25 mM)
SMILES FC(F)(F)OC1=CC=C(N2C(C)=C(C#CC3=CC(Cl)=NC=C3)N=C2C)C=C1
Molecular Weight (g/mol) 391.77
Formula Weight 391.77
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.