missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CTPI 2
GREENER_CHOICE

Product Code. 30000413
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30000413

Brand: Tocris Bioscience™ 7794/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective mitochondrial citrate transporter (SLC25A1) inhibitor

CTPI 2 is a selective mitochondrial citrate transporter (SLC25A1) inhibitor with a KD of 3.5 μM. CTPI 2 inhibits the proliferation of H1299 cells in a SLC25A1-dependent manner. It also re-sensitizes T1, T2, and T4 cells to Cisplatin (Cat. No. 2251). CTPI 2 inhibits tumor growth in in vivo models of non-small cell lung cancer (NSCLC). In high-fat diet fed mice CTPI 2 regulates glycolysis, prevents steatohepatitis and normalizes glucose tolerance and insulin sensitivity. CTPI 2 influences the inflammatory pathway by inhibiting IL-6 and TNFα production and M1 macrophage polarization.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[[(4-Chloro-3-nitrophenyl)sulfonyl]amino]benzoic acid
CAS 68003-38-3
Quantity 10 mg
Target Other Ion Pump/Transporter Inhibitors
Molecular Formula C13H9ClN2O6S
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.