missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ B-27™ Supplement, XenoFree, minus vitamin A

Product Code. 15934204
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
This item is not returnable. View return policy

Product Code. 15934204

Brand: Gibco™ A3353501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

B-27™ Supplement, XenoFree, minus vitamin A

B-27 Supplement XenoFree Minus Vitamin A is based on the classic B-27 Supplement formulation and contains no animal-derived components at the primary component level. The absence of vitamin A (retinyl acetate) makes this formulation ideal for the cultivation of neural progenitor and stem cells, either as neurospheres in suspension or in adherent monolayer culture, without inducing differentiation.

Quality

B-27 Supplement XenoFree Minus Vitamin A is manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified.

Formulated xeno-free

The preparation contains no animal-derived components at the primary component level.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -5°C to -20°C and protect from light.
Description Without Vitamin A
Cell Type Neural Progenitor Cells
Classification Serum-free
Concentration 50 X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Cell Culture Supplement
Serum Level Serum-Free
Sterility Sterile
pH 6 to 8
Product Line B-27
Quantity 10 mL
Shipping Condition Dry Ice
Source Xeno-Free
Tested For Endotoxin, Performance, Sterility, Mycoplasma
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.