Learn More
Description
B-27 Supplement XenoFree Minus Insulin is based on the classic B-27 Supplement formulation and contains no animal-derived components at the primary component level. The absence of insulin makes this formulation ideal for the growth of hippocampal and other CNS neurons in applications where the presence of insulin causes interference or in the derivation of cardiomyocytes and mesodermal cells from stem cells.
Quality
B-27 Supplement XenoFree Minus Insulin is manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified.
Formulated xeno-free
The preparation contains no animal-derived components at the primary component level.
Specifications
Specifications
| Content And Storage | Store at -5°C to -20°C and protect from light. |
| Description | Without Insulin |
| Cell Type | Neural Cells |
| Classification | Serum-free |
| Concentration | 50 X |
| Culture Type | Mammalian Cell Culture |
| Form | Liquid |
| Product Type | Cell Culture Supplement |
| Serum Level | Serum-Free |
| Sterility | Sterile |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.