missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ B-27™ Supplement, XenoFree, minus vitamin A (50X)

Product Code. 17875572 Shop All Gibco Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17875572 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17875572 Supplier Gibco™ Supplier No. A5047601
 Restricted Item
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CTS B-27 Supplement, XenoFree, minus vitamin A, is based on the classic serum-free B-27 supplement and designed to help meet regulatory requirements for cell therapy manufacturing.

Gibco CTS B-27 Supplement, XenoFree, minus vitamin A, is a cGMP-manufactured, serum-free supplement designed to meet regulatory requirements for cell therapy manufacturing. It supports the growth and differentiation of a variety of cell types derived from pluripotent stem cells (PSCs). The absence of vitamin A makes this formulation ideal for the cultivation and expansion of neural progenitor and stem cells without inducing differentiation until desired.

CTS quality

Gibco CTS products are manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified.

Accompanied by documentation to support your regulatory filing

  • Access to our Drug Master File (DMF) or Regulatory Support File (RSF)
  • Traceability documentation—Certificate of Origin, Certificate of Analysis, and Safety Data Sheet

Reduced risk from animal-origin materials and human-derived components

CTS B-27 Supplement, XenoFree, minus vitamin A, is formulated with human-derived components and AOF recombinant components. To help reduce viral clearance risks, the use of human-derived components has been minimized and human-derived components are limited to materials that help meet regulatory standards for donor qualification, viral testing, and viral inactivation.

CTS B-27 Supplement, XenoFree, minus vitamin A, was specifically designed for use in cell therapy applications by making targeted modifications to the classic serum-free B-27 Supplement. CTS B-27 Supplement, XenoFree, minus vitamin A, supports the robust growth, expansion, and differentiation of stem cells, making it a valuable addition to any cell therapy protocol where cGMP standards are required.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -5°C to -20°C and protect from light.
Classification Serum-free, Xeno-free
Concentration 50X
Culture Type Cells derived from Pluripotent Stem Cells (PSCs)
Form Liquid
Product Type Cell Culture Supplement
Sterility Sterile-filtered
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity 10 mL
Shipping Condition Dry Ice
Show More Show Less

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.