missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Detachable Dynabeads™ CD3/CD28 Kit

Product Code. 17846158
Change view
Click to view available options
Quantity:
Large Kit
Small Kit
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17846158 Small Kit Each
17806158 Large Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17846158 Supplier Gibco™ Supplier No. A56992

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Gibco CTS Detachable Dynabeads CD3/CD28 Kit is intended for ex vivo isolation and activation of T cells in a simultaneous one-step mechanism.

The Gibco CTS Detachable Dynabeads CD3/CD28 Kit is intended for ex vivo isolation and activation of T cells in a simultaneous one-step mechanism. As part of a next-generation platform, this offering is the first cGMP cell selection technology to have an active release mechanism for process development, clinical trial, and commercial manufacturing use.

Features of the CTS Detachable Dynabeads CD3/CD28 Kit include:
• Empowers users with process flexibility by allowing active release of beads from target cells at any time
• Optimizes target cell purity, yield, activation profile, and desired phenotype thanks to the active release mechanism that gives users more control
• Provides an automation-friendly solution by working directly with the CTS DynaCellect Magnetic Separation System
• Enables consistent performance in light of biological variability within processes
• Is suitable for both autologous and allogeneic cell therapy manufacturing processes

Coated with camelid-derived single-domain VHH antibodies that target CD3 and CD28 cell surface markers, these Dynabeads magnetic beads provide both the primary and co-stimulatory signals that are required for activation and expansion of T cells. An accompanying release buffer within the kit has an active ingredient, biotin, that competitively displaces a biotin derivative on the surface of the beads. This proprietary mechanism allows users to achieve active release of the beads from target cells at any time within their process.

Based on a recommended 3-to-1 bead-to-target-cell ratio (not total nucleated cell number), the smaller kit size can theoretically isolate and activate up to 1.33 x 109 T cells. The larger kit size can theoretically isolate and activate up to twice that number of cells.

The CTS Detachable Dynabeads CD3/CD28 Kit also includes Drug Master Files (DMFs) for each of its components to support cross-referencing in regulatory submissions prior to clinical trial and commercial manufacturing use.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 10 mL CTS Detachable Dynabeads CD3/CD28 (1 vial)
• 400 mL CTS Detachable Dynabeads Release Buffer (1 bioprocess container)

Store at 2–8°C.

Color Brown
Material Polystyrene
Product Type Cell Isolation and Activation Reagent
Quantity Small Kit
Diameter (Metric) 4.5 micron

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.