Learn More
Gibco™ CTS™ Detachable Dynabeads™ Release Buffer
Description
The Gibco CTS Detachable Dynabeads Release Buffer is for use with CTS Detachable Dynabeads magnetic beads. When used together, they empower users with a first-of-its-kind cGMP cell selection technology that has an active release mechanism for process development, clinical trial, and commercial manufacturing use.
Features of the CTS Detachable Dynabeads Release Buffer (when used with CTS Detachable Dynabeads magnetic beads) include:
- Empowers users with process flexibility by allowing active release of beads from target cells at any time
- Optimizes target cell purity, yield, activation profile, and desired phenotype thanks to the active release mechanism that gives users more control
- Provides an automation-friendly solution by working directly with the CTS DynaCellect Magnetic Separation System
- Enables consistent performance in light of biological variability within processes
- Is suitable for both autologous and allogeneic cell therapy manufacturing processes
The CTS Detachable Dynabeads Release Buffer has an active ingredient, biotin, that competitively displaces a biotin derivative on the surface of CTS Detachable Dynabeads magnetic beads. This proprietary mechanism allows users to achieve active release of the beads from target cells at any time within their process. And when used on the CTS DynaCellect Magnetic Separation System, these products deliver automation, flexibility, and scalability in cell therapy manufacturing workflows.
The CTS Detachable Dynabeads Release Buffer also includes Drug Master Files (DMFs) to support cross-referencing in regulatory submissions.
Specifications
Specifications
| Color | Clear |
| Concentration | 5 mM |
| For Use With (Application) | Cell Therapy |
| For Use With (Equipment) | CTS DynaCellect Magnetic Separation System |
| pH | 7.0 to 7.4 |
| Physical Form | Liquid |
| Quantity | 400 mL |
| Solution Type | Buffer |
| Grade | GMP |
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.