missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ GlutaMAX™ Supplement

Product Code. 18801544 Shop All Gibco Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Format:
Bag - BioProcess Container (BPC)
Bottle
Unit Size:
100mL
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format Unit Size
18801544 1 L Bag - BioProcess Container (BPC) Each
10860311 100 mL Bottle 100mL
17246533 100 mL Bag - BioProcess Container (BPC) 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18801544 Supplier Gibco™ Supplier No. A4737003
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CTS™ GlutaMAX™ Supplement is an animal origin free (AOF) dipeptide alternative to L-glutamine, with increased stability that improves cell health. CTS™ GlutaMAX™ Supplement can be used as a direct substitute for L-glutamine at equimolar concentrations in mammalian and stem cell culture with minimal or no adaptation.

CTS GlutaMAX Supplement is an animal origin free (AOF) dipeptide alternative to L-glutamine, with increased stability that improves cell health. CTS GlutaMAX Supplement can be used as a direct substitute for L-glutamine at equimolar concentrations in mammalian and stem cell culture with minimal or no adaptation.

Gibco CTS products are designed for use in manufacturing of cell or gene products, supporting your transition from research to clinical applications.

Compared to L-glutamine, CTS GlutaMAX Supplement:

  • Minimizes toxic ammonia build-up - Unlike L-glutamine, CTS GlutaMAX Supplement does not spontaneously break down to form ammonia. This prevents waste build-up and maintains a fresh supply of L-glutamine during long-term culture.
  • Improves growth efficiency and performance of mammalian cell cultures - Lower levels of ammonia result in improved cell viability. In addition, culture conditions are more controlled due to the consistent concentration of CTS GlutaMAX Supplement that is not possible with L-glutamine.
  • Remains stable across a wide range of temperatures - CTS GlutaMAX Supplement retains full functionality at room temperature or 2 to 8°C, for convenient storage and handling.

Closed systems compatible

This size of CTS GlutaMAX Supplement is provided in bioprocess container format with the chamber constructed from Aegis 5-14 film. Sterile, weldable C-Flex™ tubing with a female luer lock provides versatile connection options for integrating into a closed system manufacturing process.

CTS quality

The Gibco CTS product line enables you to reduce your burden in qualifying reagents during your transition from research to clinical applications.

Products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485- and ISO 9001 to certified. Gibco CTS products follow USP <1043> ancillary materials for cell, gene, and tissue-engineered products within the responsibilities applicable to a supplier.

Gibco CTS products are of high quality and are supplied with harmonized documentation such as certificates of analysis, certificates of origin, and convenient access to our drug master file (DMF).

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type Cell Culture Supplement
Sterility Sterile-filtered
Sterilization Method 0.2 μm Sterile-filtered
Format Bag - BioProcess Container (BPC)
Quantity 1 L
Shipping Condition Room Temperature

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.