missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ GlutaMAX™ Supplement
GREENER_CHOICE

Product Code. p-200105788
Click to view available options
Quantity:
100 mL
Format:
Bag - BioProcess Container (BPC)
Bottle
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 10860311

Brand: Gibco™ A1286001

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CTS™ GlutaMAX™ Supplement is an animal origin free (AOF) dipeptide alternative to L-glutamine, with increased stability that improves cell health. CTS™ GlutaMAX™ Supplement can be used as a direct substitute for L-glutamine at equimolar concentrations in mammalian and stem cell culture with minimal or no adaptation.

CTS GlutaMAX Supplement is an animal origin free (AOF) dipeptide alternative to L-glutamine, with increased stability that improves cell health. CTS GlutaMAX Supplement can be used as a direct substitute for L-glutamine at equimolar concentrations in mammalian and stem cell culture with minimal or no adaptation.

Gibco CTS products are designed for use in manufacturing of cell or gene products, supporting your transition from research to clinical applications.

Compared to L-glutamine, CTS GlutaMAX Supplement:

  • Minimizes toxic ammonia build-up - Unlike L-glutamine, CTS GlutaMAX Supplement does not spontaneously break down to form ammonia. This prevents waste build-up and maintains a fresh supply of L-glutamine during long-term culture.
  • Improves growth efficiency and performance of mammalian cell cultures - Lower levels of ammonia result in improved cell viability. In addition, culture conditions are more controlled due to the consistent concentration of CTS GlutaMAX Supplement that is not possible with L-glutamine.
  • Remains stable across a wide range of temperatures - CTS GlutaMAX Supplement retains full functionality at room temperature or 2 to 8°C, for convenient storage and handling.

Closed systems compatible

This size of CTS GlutaMAX Supplement is provided in bioprocess container format with the chamber constructed from Aegis 5-14 film. Sterile, weldable C-Flex™ tubing with a female luer lock provides versatile connection options for integrating into a closed system manufacturing process.

CTS quality

The Gibco CTS product line enables you to reduce your burden in qualifying reagents during your transition from research to clinical applications.

Products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485- and ISO 9001 to certified. Gibco CTS products follow USP <1043> ancillary materials for cell, gene, and tissue-engineered products within the responsibilities applicable to a supplier.

Gibco CTS products are of high quality and are supplied with harmonized documentation such as certificates of analysis, certificates of origin, and convenient access to our drug master file (DMF).

TRUSTED_SUSTAINABILITY

Spezifikation

Content And Storage Storage Conditions: Room Temperature
Cell Type Mammalian and stem cell culture
Classification Xeno-free
Concentration 100X
Form Liquid
Product Type Supplement
Sterility Sterile-filtered
Format Bottle
For Use With (Application) Cell and Gene Therapy Research, Development and Manufacturing
Green Features Sustainable packaging
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity 100 mL
Shipping Condition Room Temperature
Mehr anzeigen Weniger anzeigen

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt