missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ HBSS (1X)

Product Code. 30534456
Click to view available options
Quantity:
1 L
Unit Size:
1000mL
This item is not returnable. View return policy

Product Code. 30534456

Brand: Gibco™ A4000225401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gibco Cell Therapy Systems (CTS) Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

Gibco Cell Therapy Systems (CTS) Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. This particular formulation lacks calcium, magnesium and phenol red.

CTS quality

The Gibco CTS product line enables you to reduce your burden in qualifying reagents during your transition from research to clinical applications.

Products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485- and ISO 9001–certified. Gibco CTS products follow USP <1043> ancillary materials for cell, gene, and tissue-engineered products within the responsibilities applicable to a supplier.

Gibco CTS products are of high quality and are supplied with harmonized documentation such as certificates of analysis and certificates of origin.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Culture Type Mammalian and stem cell culture
Form Clear, colorless solution
Product Type Cell Culture Supplement
Sterility Sterile-filtered
Sterilization Method 0.2 μm Sterile-filtered
Format Bottle
For Use With Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing Quality ISO13485, MDSAP, FDA-registered, 21 CFR820
Quantity 1 L
Shipping Condition Ambient
Show More Show Less

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.