missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ LV-MAX™ Production Medium

Product Code. 17133189
Change view
Click to view available options
Quantity:
1 L
10 L
20 L
6 x 1 L
Unit Size:
1000mL
20 L
Each
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17133189 6 x 1 L Pack of 6
16356572 1 L 1000mL
16366572 10 L Each
16376572 20 L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17133189 Supplier Gibco™ Supplier No. A4124004
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gibco™ CTS LV-MAX Production Medium is a chemically-defined, serum-free, protein-free medium validated for recombinant lentiviral vector production.

Gibco™ CTS LV-MAX Production Medium is a chemically-defined, serum-free, protein-free medium validated for recombinant lentiviral vector production. In addition to supporting high cell density suspension cultures of CTS Viral Production Cells (HEK293F-derived cells), this medium is suitable for preclinical-, clinical-, and commercial-scale transient transfection and lentiviral production when used with the CTS LV-MAX Transfection Kit. CTS LV-MAX Production Medium undergoes stringent QC testing to ensure it is appropriate for the manufacturing of cell and gene therapy applications.

CTS LV-MAX Production Medium may be applicable for other recombinant viral vector production that uses a HEK293 medium.

CTS LV-MAX Production Medium features include:

  • Supports CTS Viral Production Cells to achieve densities of over 1 x 107 cells/mL with minimal cell clumping
  • Enables high viral titers of up to 1 x 108 TU/mL (unconcentrated lentiviral vectors–GFP)
  • Scalable protocol for 96-deep well blocks, flasks, and bioreactors
  • Regulatory-compliant medium manufactured in conformity with cGMP for medical devices, 21 CFR, Part 820, and certified to the ISO 13485. Drug Master File and Regulatory Support File available upon request.

CTS LV-MAX Production Medium is formulated with GlutaMAX-I reagent. It is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. The medium contains no human or animal-origin components. It is not recommended for adherent cell cultures. The formulation of this product is identical to research-grade LV-MAX Production Medium, but CTS LV-MAX Production Medium is validated for clinical and commercial therapeutic applications.

Custom configuration and packaging available upon request.

TRUSTED_SUSTAINABILITY

Spezifikation

Cell Line Viral Production Cells
Cell Type HEK293
Classification Animal Origin-free
Form Liquid
Product Type LV Production Medium
Sterility Sterile-filtered
For Use With (Application) LV Production
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity 6 x 1 L
Shipping Condition Ambient
Format Bottle
Content And Storage 6 x 1 L bottles. Store at 2°C to 8°C and protect from light.
Mehr anzeigen Weniger anzeigen
Are CTS Viral Production Cells 293T-derived?

No, CTS Viral Production Cells are HEK 293F suspension cells that have been adapted to CTS LV-MAX Production Medium. They do not express the SV40 large T antigen.

How do I bank CTS Viral Production Cells?

CTS Viral Production Cells can be frozen directly in cryogenic medium consisting of 90% CTS LV-MAX Production Medium plus 10% DMSO. Here is the protocol:

- Bank cells at passage 3 when cell density reaches ˜3.0-4.5 x 10E6 viable cells/mL at >95% viability.
- Centrifuge the cells at 100 x g (low speed) for 5 mins to pellet. Discard the spent medium.
- Prepare freezing medium: e.g., 10 mL of freezing medium is 9 mL of CTS LV-MAX Production Medium + 1 mL DMSO; mix well and sterilize by 0.2 µm filtration.
- Resuspend the cell pellet in the prepared freezing medium.
- Dilute the cells to a final density of 10 x10E6 viable cells/mL and aliquot 1 mL per cryovial.
- Freeze the cells at -80 degrees C for 1 day.
- Transfer frozen vials to liquid nitrogen for long-term storage.

Why do you recommend using V bottom 96-well plates for harvesting lentivirus from CTS Viral Production Cells?

We recommend using V-bottom 96-well plates to harvest the supernatant containing lentiviral particles as they allow for easier detection of the cell pellets after centrifugation of the cultures.

During lentivirus production from CTS Viral Production Cells, will the shaker vibration affect the cell growth of other cultures maintained in the same incubator or in stacked incubators?

We recommend maintaining shaking cultures in a separate incubator from all other incubators to avoid vibration effects. Alternatively, we recommend using a small, quiet shaker such as the Thermo Scientific CO2 Resistant Shaker (Cat. No. 88881102) for convenience and best performance.

Is 8% CO2 a growth requirement for CTS Viral Production Cells?

We generally recommend culturing the CTS Viral Production Cells under 8% CO2 conditions. We found that cells cultured in 5% CO2 grew slower and were more prone to more cell clumping than when grown in 8% CO2.

What is special about the CTS LV-MAX lentivirus production system?

The CTS LV-MAX lentivirus production system is a fully-optimized complete system in which the cells, media, and transfection reagent work together to generate high-titer lentivirus. The CTS LV-MAX Production Medium is specifically designed to work with the system, to support high density suspension cell growth of CTS Viral Production Cells.

The manual recommends using CTS Viral Production Cells passaged to P5 (˜3 weeks) on the day of transfection. Are these general cell culture guidelines or special requirements for these cells?

Although 3-4 passages before transfection are sufficient for most cell lines recovering directly from liquid N2 storage, we found that CTS Virus Production Cells passaged to P5 showed the highest performance in lentivirus particle production and reproducibility between batches. Therefore, we recommend passaging the cells to P4 at the end of a given week for lentivirus production the following week.

The manual recommends passaging the CTS Viral Production Cells three times before freezing. Can I bank the cells sooner than that?

For optimal cell recovery from liquid N2 storage, CTS Viral Production Cells should be passaged to passage 2 (P2) (from the original vial) and up to P3 before banking, to ensure good cell health and viability. Cells that demonstrate good cell health (i.e., ˜26 hr doubling time, round cell morphology, and minimal clumping) may be banked at P2 or P3.

The CTS LV-MAX lentivirus production system user manual recommends the passage of CTS Viral Production Cells every 3-4 days; is this necessary? Can I seed the cells less densely and passage less frequently (i.e., every 6-7 days)?

Passaging the CTS Viral Production Cells every 3-4 days allows for cell growth rates to progress from lag to log phase, thus ensuring highest cell viability after splitting cultures. We recommend passaging the cells at a density of no lower than ˜0.35x10E6 viable cells/mL as low cell densities maintained in the lag phase for long periods, will eventually lead to poor overall cell growth. We do not recommend culturing the cells past 4 days.
The cell culture densities for CTS Viral Production Cells are as follows:
- 0.35x10E6 and 0.55x10E6 viable cells/mL (splitting cell density for 4 day and 3 day culture, respectively)
- 3.5x10E6 to 5x10E6 viable cells/mL (subculture cell density)
- Log-phase is from 0.35x10E6 to 6.5x10E6 viable cells/mL.

With the CTS LV-MAX lentivirus production system, do you have any recommendations for how I can produce more lentiviral expression plasmid?

When propagating lentiviral expression plasmids, it is important to propagate them in cells that will minimize any sequence rearrangements or unwanted recombination due to the long terminal repeat (LTRs) sequences and large size of the vector backbone ( ˜10 Kb). To minimize these events, we recommend using Stbl3 cells for stable propagation of lentiviral expression plasmids.

With the CTS LV-MAX lentivirus production system, I would like to titer my lentivirus using a different method from GFP or antibiotic method, is this okay?

We recommend using either the GFP method or antibiotic-based method described in the manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018450_CTS_LV-MAX_LentiviralProdSys_UG.pdf). You may try other viral titering methods, however, we do not have protocols for methods other than by GFP or antibiotics.

I am using the CTS LV-MAX lentivirus production system and my lentiviral titer is low. How do I troubleshoot effectively and what can I do to optimize this?

Low lentiviral titer could be due to many different factors such as non-optimized lentiviral expression vector design, low transfection efficiency, incorrect cell density, suboptimal culture conditions (e.g., incubator, shaker speed, vessel type, media contamination), purification strategy, and titer method. To best track your workflow from culturing your cells to titering the lentivirus, we recommend using a positive control such as the Vivid Colors pLenti6.3/V5-GW/EmGFP Expression Control Vector (Cat. No. V37006).

I am using CTS Viral Production Cells and am seeing many dead cells in my culture. What could be causing this?

This could mean that the seeding or starting cell density was too low. The CTS LV-MAX Supplement relies on high cell density. Refer to the CTS LV-MAX Lentiviral Production System manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018450_CTS_LV-MAX_LentiviralProdSys_UG.pdf) for cell density recommendations.

My CTS Viral Production Cells are clumping. Can I use an anti-clumping reagent?

Anti-clumping reagents could potentially inhibit transfection efficiency and are generally not recommended. If needed, add no more than 0.1% Pluronic F-68

Can I use other plasmids with the CTS LV-MAX lentivirus production system?

The CTS LV-MAX lentivirus production system is designed to be compatible with any format of plasmid DNA. The quality of plasmids (e.g., purity), ratio of the plasmids, and vector design all contribute to lentiviral titer. Please contact us at techsupport@thermofisher.com for suggestions.

In the CTS LV-MAX transfection protocol, CTS LV-MAX Enhancer addition time is suggested to be anytime between 5-14 hr after transfection. How critical is it that I stay within this time window?

The formulation of the CTS LV-MAX Enhancer coupled with the broad window of enhancer addition time was specifically optimized to allow user flexibility in schedule and start time of the protocol without affecting performance. For optimal results, we recommend adhering to the suggested time window for enhancer addition.

In the CTS LV-MAX protocol, the cells remain in the same media throughout the two-day transfection. Why is no media change needed?

The CTS LV-MAX Production Medium is specially designed to support the health of Viral Production Cells cultured at high-density. We have performed comprehensive studies in-house and have determined that there is no need for media change during the protocol.

Do you have recommendations for measuring CTS lentiviral titer using a non-fluorescent method?

Please refer to page 31 of the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018450_CTS_LV-MAX_LentiviralProdSys_UG.pdf) for a protocol for lentiviral titer measurement using antibiotic selection.

Are the sequences of the LV-MAX Lentiviral Packaging Mix plasmids confidential?

The sequences of the LV-MAX Lentiviral Packaging Mix plasmids are published on our website; however, the ratio of the plasmids in the mix is confidential.

What is the recommended ratio of plasmids to use in the CTS LV-MAX lentivirus production system?

We recommend using the lentiviral packaging plasmids and transfer plasmids at a ratio of 3:2.

Should I harvest the lentivirus from the CTS Viral Production Cells at 24 hr and 48 hr time-points. How about waiting for 72 hr?

Our system has been optimized to provide maximal lentiviral production at 48 hr post transfection. Waiting until 72 hr is not recommended since existing lentiviral particles within the media will infect the production cells and can reduce the titer.

The CTS Viral Production Cells are supposed to be good in passage 5-20. Does this refer to passage 5-20 post-thaw, or should we consider the number of passages the cells went through before cryopreservation?

The passages should be counted upon thawing, and the cells should be discarded when they reach passage 20 post-thaw.

Can I transfect CTS Viral Production Cells before passage 5 (P5)?

We recommend that CTS Viral Production Cells be used for lentiviral production only following passage 5 to allow for full recovery of the cells from the cryopreservation; CTS Viral Production Cells are sensitive upon thawing and need the extra passages for recovery.

Why do I need to dilute the CTS Viral Production Cells once more before transfection?

This is a necessary step to replenish nutrients and ensure that the cells are at the correct density to achieve the concentration needed for optimal transfection efficiency. It is crucial that the cells are at the proper cell density during the time of transfection to ensure maximal performance.

What density should I be seeding the CTS Viral Production Cells at?

Depending on whether your culture will be 3 or 4 days, we have outlined the differences in seeding densities in the product manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018450_CTS_LV-MAX_LentiviralProdSys_UG.pdf). We also have specific guidance for seeding densities to be used for lentiviral vector production.

Do you have protocols you can share for scale-up with the CTS LV-MAX lentivirus production system? What scale have you tested so far?

We continually develop large-scale protocols to support clinical and commercial lentiviral vector production. Currently, we have a 2 L bio-reactor protocol that is available from techsupport@thermofisher.com. We are also working on a 5 L and 50 L scale.

My CTS Viral Production Cells have low viability at 3 days post-thawing. Is this normal?

It is okay to see variation in viability upon thawing, but this should normalize during subsequent passages. We recommend culturing the cells as described in chapter 2 of the manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018450_CTS_LV-MAX_LentiviralProdSys_UG.pdf) so that the cells recover sufficiently and achieve >90% viability before starting the protocol for CTS lentviral production.

Can I use other media like HEK293 expression media with the CTS LV-MAX products?

We have tried other media and have seen reduced performance. For maximal lentiviral vector yield, we recommend using the CTS LV-MAX Production Medium.

Can I use other commercially available transfection reagents with the CTS LV-MAX Production Medium and CTS Viral Production Cells?

The CTS LV-MAX Transfection Kit is specifically designed to work synergistically with the CTS LV-MAX Production Medium and CTS Viral Production Cells to provide maximal lentiviral titer. Mixing and matching reagents is not recommended, as this will decrease the overall performance.

I'm interested only in the CTS LV-MAX Transfection Reagent. Can I purchase it separately from the CTS LV-MAX Transfection Kit?

The CTS LV-MAX Transfection Kit contains three components, the transfection reagent, supplement, and enhancer, that are not sold as stand-alone items. The system components were specifically optimized to work as a complete system to obtain maximal lentiviral titer. We can however customize different ratios of the transfection reagent, supplement, and enhancer - for further information, please send an email to customorders@thermofisher.com.

Do you have a protocol to detect residual components of the CTS LV-MAX transfection reagent and enhancer?

We can provide protocol guidance for how to detect transfection reagent components under CDA. Please contact our outlicensing@thermofisher.com for information.

Do you have any guidelines for purification of lentivirus from CTS Viral Production Cells under serum-free conditions?

For downstream purification of lentivirus under serum-free conditions, we recommend using tangential flow filtration. Please contact techsupport@thermofisher.com for the suggested protocol. In order to maintain the integrity of lentiviral vectors, we do not recommend using ultracentrifugation under serum-free conditions.

What is the difference between Expi293F Cells, Viral Production Cells, CTS Viral Production Cells, and FreeStyle 293-F Cells?

The intended use, validation, and protocol optimization varies among the different cell types. Expi293F Cells and FreeStyle 293-F Cells were specifically developed and validated for transient protein production while Viral Production Cells and CTS Viral Production Cells were designed and validated for lentiviral vector manufacturing. The commonality between the Expi293F Cells, Viral Production Cells, and CTS Viral Production Cells is that they were originally derived from FreeStyle 293-F lineage. Both Expi293F and Freestyle 293F cells come with regulatory support intended for bio-production applications whereas CTS Viral Production Cells (cGMP banked) are intended for cell and gene therapy applications, and complemented with a more robust regulatory support package to enable clinical gene therapeutic applications. This means that to the best of our ability, we keep our CTS product specifications and release requirements up-to-date in accordance with the latest regulatory guidance specific for this intended use.

What are the major differences between LV-MAX and CTS LV- MAX products?

The LV-MAX and CTS LV- MAX products have equivalent performance. In contrast to the research use only LV-MAX products, CTS LV-MAX products have an additional intended use for cell and gene therapy applications, and are complemented with a more robust regulatory support package to enable clinical therapeutic applications. This means that to the best of our ability, we keep our CTS product specifications and release requirements up-to-date in accordance with the latest regulatory guidance specific for this intended use.

What is the intended use claim for your LV-MAX and CTS LV-MAX products?

Our LV-MAX products are intended for research use only while our CTS LV- MAX products are intended for research use or manufacture of cell, gene or tissue based products. Please refer to the individual products for further details on the intended use claim.

What kind of documentation do you provide with the CTS Viral Production Cells?

We have extensive documentation to demonstrate lineage history, viral clearance, and traceability of our GMP manufactured process. Under CDA, we can share our Certificate of Analysis and Table of Contents. Full access to the documentation package is available through outlicensing@thermofisher.com.

Can I use the CTS LV-MAX products for clinical trials or commercial use?

Please refer to our standard sales terms and conditions and limited use label license. For further inquiries, please send an email to outlicensing@thermofisher.com.


For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.