missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ NK-Xpander™ Medium

Product Code. 31369470
Change view
Click to view available options
Quantity:
500 mL
5 L
10 L
20 L
Format:
BPC
BPC, Bagged Supplement (100 mL)
Bottle
Unit Size:
5 L
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format unitSize
31369470 10 L BPC -
17528106 500 mL Bottle 500mL
17661536 5 L BPC 5 L
31369778 5 L BPC, Bagged Supplement (100 mL) -
31370164 20 L BPC -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31369470 Supplier Gibco™ Supplier No. A5019003
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

CTS NK-Xpander Medium is specifically formulated for superior expansion of human natural killer (hNK) cells for cell therapy applications.

Gibco CTS NK-Xpander Medium is specifically formulated for superior expansion of human natural killer (hNK) cells for cell therapy applications. When hNK cells are enriched from human peripheral blood mononuclear cells (hPBMCs) and expanded in a feeder-free system using CTS NK-Xpander Medium supplemented with 5% hAB serum and 500U/mL IL-2, ≥90% of the hNK cells generated maintain CD56+/CD16+/CD3- surface marker expression. Furthermore, such hNK cells demonstrate degranulation and killing of K562 cancer cells when expanded in CTS NK-Xpander Medium.

Superior expansion potential

CTS NK-Xpander Medium is capable of producing a high yield of hNK cells without the need for feeder cells, providing robust expansion of enriched hNK cells from hPBMCs from qualified donors at day 14 to 15 versus other medium options. Cells maintain CD56+/CD16+/CD3- surface marker expression and demonstrate cytolytic capabilities when expanded in CTS NK-Xpander Medium.

Manufactured for cell therapy applications

CTS NK-Xpander Medium is manufactured without cytokines and growth factors to allow flexibility with regard to intended application. The medium does not include human or animal-derived components; however, supplementation with hAB serum is recommended for best results.

CTS quality

Gibco CTS products follow USP <1043> ancillary materials for cell, gene, and tissue-engineered products within the responsibilities applicable to a supplier.* Documentation such as specific intended-use statements, full documentation traceability, and convenient access to our Drug Master File (DMF) are available to support your regulatory filing.

The products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485-certified.

Closed-system compatibility

CTS NK-Xpander Medium is available in 500-mL-bottle and 5-L-bag formats for convenient process development and closed-system manufacturing scale-up. The 5-L bioprocess container chamber is constructed from Aegis 5-14 film and includes sterile, weldable DEHP-free PVC tubing with Luer lock and C-Flex™ tubing with MPC quick connector providing versatile connection options for your manufacturing process, including compatibility with the CTS Rotea Counterflow Centrifugation System.

*Other aspects of USP <1043> will be the responsibility of the end-user to assess. Thermo Fisher Scientific cannot fulfill USP <1043> in regards to application and therapy-specific aspects (e.g., residual assessment and removal of the AM).

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Natural Killer (NK) cells
Classification Animal Origin
Form Liquid
Product Type Cell Culture Medium
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
Without Additives No Phenol Red
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity 10 L
Shipping Condition Basal: Ambient, Supplement: Dry Ice
Format BPC
Content And Storage • Basal Medium: Store at 2°C to 8°C and protect from light.
• Supplement: Store at -20°C and protect from light.
Culture Type Mammalian Cell Culture
Show More Show Less
Can I use CTS NK-Xpander Medium in a feeder culture system?

Yes. Customer data have shown that CTS NK-Xpander Medium is capable of proliferating NK cells in feeder culture systems; however, as it has not been tested for use with feeder cells in-house, we recommend optimization for best results.

Can I use CTS NK-Xpander Medium to expand NK cells in a serum-free culture system?

Yes. Customer data has shown that CTS NK-Xpander Medium is capable of proliferating NK cells using 5% CTS Immune Cell Serum Replacement (SR) (Cat. No. A2596101, A2596102) with modified K562 feeder cells; however, this protocol has not been optimized in-house.

Have you tried supplementing CTS NK-Xpander Medium with IL-15, IL-21, or other cytokines?

Yes. Combinations of IL-2, IL-15, and IL-21 were tested with CTS NK-Xpander Medium; however, alternative combinations were not found to affect performance or functionality. We recommend supplementation with IL-2 only.

Does CTS NK-Xpander Medium require activation beads for NK cell proliferation?

No. Unlike cytotoxic T cells, activation beads are not required for NK cell proliferation.

Does CTS NK-Xpander Medium contain blood-derived components?

No. CTS NK-Xpander Medium is manufactured under animal origin-free (AOF) conditions; however, we recommend supplementation of CTS NK-Xpander Medium with human AB serum for best results.

Is it possible to get alternative volumes of CTS NK-Xpander Medium?

Yes. We offer 10 L and 20 L volumes (Cat. Nos. A5019003, A5019004). Custom batches of cGMP-manufactured CTS NK-Xpander Medium can be obtained in alternative bag sizes or tubing configurations through our Custom Media Service channel (custommedia@thermofisher.com).

How can I address cell attachment in my NK cell culture?

Cell attachment is more frequently seen in flasks than plates. A cell scraper can be used to gently remove cells that lightly adhere to the flask surface. We recommend using non-tissue culture treated plates and flasks when expanding NK cells.

When should I expect to see the most expansion of my NK cells using CTS NK-Xpander Medium?

The most NK cell expansion can be seen after Day 5.

Is it normal to see solute in CTS NK-Xpander Supplement?

The appearance of solute in CTS NK-Xpander Supplement is normal after thaw and does not affect performance. Once the supplement reaches ambient temperature, mix gently by inverting the bottle 8-10 times. After mixing, no solute should be visible.

At what temperature should I store CTS NK-Xpander Basal Medium and CTS NK-Xpander Supplement (50X)?

We recommend storing CTS NK-Xpander Basal Medium at 2 to 8 degrees C, protected from light and CTS NK-Xpander Supplement (50X) at -20 to -5 degrees C, protected from light.

What is the shelf life for CTS NK-Xpander Basal Medium?

The shelf-life for CTS NK-Xpander Basal Medium is 12 months when stored at 2 to 8 degrees C, protected from light. Note: Shelf-life duration is determined from Date of Manufacture as stated in the product manual. The expiration date is listed on the bottle.

What is the shelf life for CTS NK-Xpander Supplement (50X)?

The shelf-life for CTS NK-Xpander Supplement (50X) is 12 months when stored at -20 to -5 degrees C, protected from light. Note: Shelf-life duration is determined from Date of Manufacture as stated in the product manual. The expiration date is listed on the bottle.

For how long can CTS NK-Xpander complete medium be used?

CTS NK-Xpander complete medium (CTS NK-Xpander Basal Medium, CTS NK-Xpander Supplement (50X), and 5% hAB serum) is stable for 3 weeks when stored in the dark at 2 to 8 degrees C.

Does the 100 mL CTS NK-Xpander Supplement (50X) also come in a bag?

Yes. The 100 mL CTS NK-Xpander Supplement (50X) is available in a bag as part of Cat. No. A5019005 and is close system compatible.

What medium do you recommend for the expansion of Natural Killer (NK) cells?

We recommend CTS NK-Xpander Medium (Cat. Nos. A5019001, A5019002, A5019005, A5019004, A5019003) for the expansion of Natural Killer (NK) cells.

What is CTS?

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.


For Research Use or Manufacturing of Cell, Gene, or Tissue- Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.