missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ StemPro™-34 SFM XF Medium

Product Code. 30565075 Shop All Gibco Products
Change view
Click to view available options
Quantity:
500 mL
2 L
Product Type:
BPC Kit
Bottle Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type
30565075 500 mL Bottle Kit
30565079 2 L BPC Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30565075 Supplier Gibco™ Supplier No. A6636901
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gibco CTS StemPro-34 SF XenoFree Medium is a flexible, optimized medium for hematopoietic stem and progenitor cell applications.

Gibco CTS StemPro-34 SF XenoFree Medium is a highly effective medium designed for the growth and development of human hematopoietic cells in culture, including hematopoietic stem cells (HSCs) and progenitor cells sourced from bone marrow, neonatal cord blood, and peripheral blood.

Key benefits for cell and gene therapy applications

  • Serum-free and xeno-free—formulated without serum or non-human derived components to comply with global regulatory standards for ancillary materials in cell therapy manufacturing; formulated with L-glutamine and without phenol red
  • Supports cell differentiation—facilitates differentiation of induced pluripotent stem cells (iPSCs) to HSCs and further differentiation into immune cells (NK cells, T-cells, B-cells, macrophages, etc.)

Regulatory compliance and quality assurance

  • cGMP manufactured—produced in accordance with 21 CFR part 820 and ISO 9001 and ISO 13485 standards to help ensure the highest quality and safety
  • USP <1043> compliant—designed to meet USP <1043> standards for ancillary materials used in cell, gene, and tissue-engineered products

Seamless integration with closed system workflows

CTS StemPro-34 SF XenoFree Medium is available in both 500-mL bottle and 2-liter bioprocessing container (BPC) formats, supporting processes through cell therapy development and manufacturing. The BPC chamber is constructed from Aegis 5-14 film and includes sterile, weldable DEHP-free PVC tubing with Luer lock and C-Flex tubing with MPC quick connector. This helps provide versatile connection options, helping ensure maximum sterility and efficiency in your manufacturing process. The 2-L kit format includes both supplement and basal components in BPCs for closed system workflows.

Comprehensive documentation for regulatory support

To facilitate your regulatory submissions, CTS StemPro-34 SF XenoFree Medium comes with an extensive documentation package, including certificates of analysis (COA), access to drug master files (DMFs), and/or regulatory support files (RSFs), and certificates of origin (COO) upon request.

Choose CTS StemPro-34 SF XenoFree Medium for a robust, regulatory-compliant solution that supports the critical processes in cell and gene therapy manufacturing, helping ensure the highest standards of quality and safety.

TRUSTED_SUSTAINABILITY

Specifications

Classification Xeno-free, Serum-free, cGMP
Form Liquid
Product Type Bottle Kit
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Glutamine
Without Additives No Phenol Red
Quantity 500 mL
Culture Type Stem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic), Suspension Cell Culture, 3D Cell Culture

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.