missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Versene™ Solution

Product Code. 16282562
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16282562 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16282562 Supplier Gibco™ Supplier No. A4239101
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

CTS Versene Solution is a cGMP-manufactured EDTA solution for use as a gentle non-enzymatic cell dissociation reagent.

CTS Versene Solution is a cGMP-manufactured EDTA solution for use as a gentle non-enzymatic cell dissociation reagent. It is formulated as 0.2 g EDTA(Na4) per liter of phosphate buffered saline (PBS) (0.48 mM). It reduces the burden in qualifying reagents when transitioning from the research bench to the clinic.

Maintain viability over multiple passages

PSCs passaged with this cGMP manufactured EDTA dissociation reagent maintain viability.

Designed with transition in mind

CTS Versene Solution has the same formulation as RUO Versene Solution but with additional characterization and quality control testing. CTS Versene Solution is manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing site is ISO 13485-certified. Full documentation traceability is available for CTS Versene Solution.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2–8°C and protect from light

Cell Type Pluripotent Stem Cells (PSCs)
Classification Animal Origin-free
Concentration 1X
Form Liquid
Product Type Cell Culture Dissociation Reagent
Sterility Sterile-filtered
Format Bottle
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity 100 mL
Shipping Condition Room Temperature
Show More Show Less

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.