missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Cultrex Rat Collagen I
Description
Tested for the ability to promote cell attachment and spreading of HT-1080 human fibro-sarcoma cells.
Order Info
The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below
Specifications
Specifications
| Content And Storage | Product is stable for a minimum of 3 months if stored at 4°C |
| Product Type | Collagen |
| Endotoxin Level | <0.20EU per 1μg of the protein by the LAL method |
| Formulation | Type I Collagen provided at 5mg/mL (Sircol Assay). Sterility Testing: No bacterial or fungal growth detected after incubation at 37°C for 14 days following USP XXIV Chapter 71 sterility test. No mycoplasma contamination detected by PCR. |
| For Use With (Application) | Bioactivity |
| pH | 3.4 to 3.6 |
| Quantity | 100 mg |
| Regulatory Status | RUO |
| Source | Rat tail tendons |
For research use only. Not for use in diagnostic procedures
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?