missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cultrex Stem Cell Qualified RGF Basement Membrane Extract

Product Code. 18472400
Change view
Click to view available options
Quantity:
1 mL
5 mL
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18472400 5 mL 5mL
18452530 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18472400 Supplier R&D Systems™ Supplier No. 343400502
  • ONLINE EXCLUSIVE PRICE
    351.00 €
    331.80 € / 5mL
    Save 19.20 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applications: Bioactivity

Promotes the attachment of human iPSCs. Effectively maintains human iPSCs in a pluripotent state in a feeder-free culture.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 8-12 mg/mL (by Lowry assay)
For Use With (Application) Bioactivity
Product Line R&D Systems
Quantity 5 mL
Endotoxin Level ≤ 8EU/mL
Source Murine Engelbreth-Holm-Swarm (EHS) tumor.
Content And Storage Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.