missing translation for 'onlineSavingsMsg'
Learn More

Cumylamine, TRC

Product Code. 30359560
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30359560 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30359560 Supplier Toronto Research Chemicals Supplier No. TRCC8347005G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cumylamine
CAS 585-32-0
Molecular Formula C9 H13 N
Quantity 5 g
Synonym Benzenemethanamine, α,α-dimethyl-, Benzylamine, α,α-dimethyl- (8CI), α,α-Dimethylbenzenemethanamine, (2-Phenyl-2-propyl)amine, 1-Methyl-1-phenylethylamine, 1-Phenylisopropylamine, 2-Amino-2-phenylpropane, 2-Phenyl-2-propanamine, Cumylamine, NSC 66156, α,α-Dimethylbenzylamine
InChI Formula InChI=1S/C9H13N/c1-9(2,10)8-6-4-3-5-7-8/h3-7H,10H2,1-2H3
SMILES CC(C)(N)c1ccccc1
IUPAC Name 2-phenylpropan-2-amine
Molecular Weight (g/mol) 135.21
Formula Weight 135.1048
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.