missing translation for 'onlineSavingsMsg'
Learn More

Cupric Sulfate Pentahydrate, ACS Reagent Grade, Ricca Chemical

Product Code. 31750361
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
Unit Size:
125g
2.50 L
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31750361 2.5 kg 2.50 L
31750135 125 g 125g
31750212 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31750361 Supplier Ricca Chemical Company Supplier No. RDCC09002.5A5
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 102
Physical Form Fine Crystals or Crystalline Powder
Quantity 2.5 kg
Packaging Natural Poly Bottle
Percent Purity 98 to 102%
Grade ACS
Chemical Name or Material Cupric Sulfate Pentahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.