missing translation for 'onlineSavingsMsg'
Learn More

Curdione, MedChemExpress

Product Code. 16474817
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474817 50 mg 50mg
16408958 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16474817 Supplier Medchem Express Supplier No. HYN0353
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Curdione, one of the major sesquiterpene compounds from Rhizoma Curcumae, has been shown to exhibit multiple bioactive properties.

Curdione, one of the major sesquiterpene compounds from Rhizoma Curcumae, has been shown to exhibit multiple bioactive properties.

Specifications

Chemical Name or Material Curdione
CAS 13657-68-6
Color White
Purity Grade Notes Research
Molecular Formula C15H24O2
Quantity 50 mg
Synonym (+)-Curdione
Solubility Information DMSO : ≥ 100 mg/mL (423.10 mM)
SMILES O=C1C[C@@H](C(C)C)C(C/C(C)=C/CC[C@@H]1C)=O
Molecular Weight (g/mol) 236.35
Formula Weight 236.35
Percent Purity 98.61%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.